Please login to your online account to display your discounted pricing

Alfa Aesar™ VERTEC™ LAC1000

$75.73 - $951.54

Chemical Identifiers

Chemical Name or Material VERTEC™ LAC1000
CAS 17476-13-0
Molecular Formula C10H20N2O4Ti
Formula Weight 280.15g/mol
MDL Number MFCD11113032
Synonym [2,2',2'',2'''-(Ethanediyldinitrilo)tetrakis(ethoxy)-N,N',O,O',O'',O''']titanium(IV); N,N,N',N'-Tetrakis(2-ethoxy)ethylediamine titanium(IV)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3106614

 
alfa aesar™
H31066-14
25g Each for $305.04

AAH3106606

 
alfa aesar™
H31066-06
5g Each for $75.73

AAH3106622

 
alfa aesar™
H31066-22
100g Each for $951.54
Description & Specifications

Specifications

Quantity 25g
Quantity 25g
Chemical Name or Material VERTEC™ LAC1000
CAS 17476-13-0
Molecular Formula C10H20N2O4Ti
Formula Weight 280.15g/mol
MDL Number MFCD11113032
Synonym [2,2',2'',2'''-(Ethanediyldinitrilo)tetrakis(ethoxy)-N,N',O,O',O'',O''']titanium(IV); N,N,N',N'-Tetrakis(2-ethoxy)ethylediamine titanium(IV)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.