Thermo Scientific™ VersiPlate, 96-well PCR Plates & Frames
Flexible 96-well plates of low profile strip tubes for use in PCR and qPCR applications.
$186.52 - $186.52
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AB1805 Thermo Scientific™
AB1805
25 Plates Case for $186.52
Specifications
Description
The Thermo Scientific VersiPlates are unique, with each strip of eight tubes being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.
Highlights
  • Easier sample handling as multiple strips of samples can be handled simultaneously
  • Reduced consumables waste as only the required numbers of strips need to be removed with the remainder being stored for the next experiment
  • Can be used in instruments with or without a VersiFrame
  • The reusable polycarbonate frame can serve as a useful rack for a VersiPlate or any number of low profile strip tubes and facilitates sample loading into thermal cyclers
  • The low profile format of the VersiPlate has a lower dead volume which reduces sample evaporation
  • Available in white which improves signal reflection, reduces variation and increases the sensitivity of qPCR assays
  • Can be used with all Thermo Scientific VersiCap™ Mats, cap strips, adhesive and heat seals
Applications
  • PCR
  • qPCR/Real-Time PCR