missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Thulium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010324500N
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Natural Poly Bottle | |
| 95.93%,1.96%,0.11% | |
| 1 | |
| O3Tm2 | |
| Approximately 0°C | |
| ZIKATJAYWZUJPY-UHFFFAOYSA-N | |
| dithulium(3+) trioxidandiide | |
| 500 mL |
| 12036-44-1,7697-37-2,12036-44-1 | |
| 99.85%,2.04%,0.11% | |
| Liquid | |
| MFCD00011285 | |
| Standard | |
| [O--].[O--].[O--].[Tm+3].[Tm+3] | |
| 385.87 | |
| High Purity |
Chemical Identifiers
| 12036-44-1 | |
| 385.87 | |
| ZIKATJAYWZUJPY-UHFFFAOYSA-N | |
| [O--].[O--].[O--].[Tm+3].[Tm+3] |
| O3Tm2 | |
| MFCD00011285 | |
| dithulium(3+) trioxidandiide |