missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Strontium (Sr) Standard for AAS 1000 ppm in 2% HCl, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

$220.66 - $388.69

Chemical Identifiers

CAS 10476-85-4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RV010053100
View Documents
Ricca Chemical Company
RV010053100N
100 mL
Each for $220.66
Only null left
 
RV010053500
View Documents
Ricca Chemical Company
RV010053500N
500 mL
Each for $388.69
Only null left
 
Description

Description

  • 1000 ppm in 2% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Chemical Identifiers

10476-85-4

Specifications

Natural LDPE Bottle
95.86%,1.96%,0.18%
1.01
Approximately 0°C
High Purity
10476-85-4,7647-01-0,7732-18-5
99.78%,2.04%,0.18%
Liquid
100 mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'