missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

VeriSpec™ Cesium Standard for ICP, 1000ppm in H2O, ISO 17025/Guide 34, Ricca Chemical

Formulated for high performance and reliable results

$170.73 - $791.67

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.355
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name cesium;chloride
SMILES [Cl-].[Cs+]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RV01027450N
View Documents Promotion Details
Ricca Chemical Company
RV01027450N
50mL
Each for $170.73
 
RV010274100
View Documents Promotion Details
Ricca Chemical Company
RV010274100N
100mL
Each for $304.73
 
RV010274250
View Documents Promotion Details
Ricca Chemical Company
RV010274250N
250mL
Each for $507.97
 
RV010274500
View Documents Promotion Details
Ricca Chemical Company
RV010274500N
500mL
Each for $791.67
 
Description

Description

  • 1000ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Chemical Identifiers

7647-17-8
168.355
24293
cesium;chloride
ClCs
AIYUHDOJVYHVIT-UHFFFAOYSA-M
CHEBI:63039
[Cl-].[Cs+]

Specifications

7647-17-8,7732-18-5
100%,0.1326%
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium;chloride
24293
Liquid
Bottle, Poly Natural
1
97.8726%,0.1274%
ClCs
[Cl-].[Cs+]
168.355
CHEBI:63039
High Purity
50mL
SDS
Product Certifications