missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Veratrole, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 161110010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Veratrole | |
91-16-7 | |
206°C to 207°C | |
Authentic | |
Glass bottle | |
1.5330 to 1.535 | |
MFCD00008357 | |
06, 771 | |
15, 10150 | |
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,methanol,acetone and ether | |
COC1=CC=CC=C1OC | |
138.17 | |
CHEBI:59114 | |
99+% |
99+% | |
1.0800g/mL | |
87°C | |
99% min. (GC) | |
C8H10O2 | |
C6H4(OCH3)2 | |
1kg | |
1.08 | |
veratrole, veratrol, pyrocatechol dimethyl ether, o-dimethoxybenzene, catechol dimethyl ether, benzene, 1,2-dimethoxy, 2-methoxyanisole, o,o-dimethyl catechol, 2-dimethoxybenzol, benzene, o-dimethoxy | |
ABDKAPXRBAPSQN-UHFFFAOYSA-N | |
1,2-dimethoxybenzene | |
7043 | |
138.17 |
Chemical Identifiers
91-16-7 | |
138.17 | |
ABDKAPXRBAPSQN-UHFFFAOYSA-N | |
7043 | |
1,2-dimethoxybenzene |
C8H10O2 | |
MFCD00008357 | |
veratrole, veratrol, pyrocatechol dimethyl ether, o-dimethoxybenzene, catechol dimethyl ether, benzene, 1,2-dimethoxy, 2-methoxyanisole, o,o-dimethyl catechol, 2-dimethoxybenzol, benzene, o-dimethoxy | |
CHEBI:59114 | |
COC1=CC=CC=C1OC |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 202-045-3
RTECSNumber : CZ6475000
TSCA : TSCA
RUO â Research Use Only