missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Veratrole, 99+%, Thermo Scientific™
Chemical Identifiers
CAS | 91-16-7 |
---|---|
Molecular Formula | C8H10O2 |
Molecular Weight (g/mol) | 138.17 |
MDL Number | MFCD00008357 |
InChI Key | ABDKAPXRBAPSQN-UHFFFAOYSA-N |
Synonym | veratrole, veratrol, pyrocatechol dimethyl ether, o-dimethoxybenzene, catechol dimethyl ether, benzene, 1,2-dimethoxy, 2-methoxyanisole, o,o-dimethyl catechol, 2-dimethoxybenzol, benzene, o-dimethoxy |
PubChem CID | 7043 |
ChEBI | CHEBI:59114 |
IUPAC Name | 1,2-dimethoxybenzene |
SMILES | COC1=CC=CC=C1OC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC161111000
|
Thermo Scientific Chemicals
161111000 |
100g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC161115000
|
Thermo Scientific Chemicals
161115000 |
500g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC161110010
|
Thermo Scientific Chemicals
161110010 |
1kg | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
91-16-7 | |
138.17 | |
ABDKAPXRBAPSQN-UHFFFAOYSA-N | |
7043 | |
1,2-dimethoxybenzene |
C8H10O2 | |
MFCD00008357 | |
veratrole, veratrol, pyrocatechol dimethyl ether, o-dimethoxybenzene, catechol dimethyl ether, benzene, 1,2-dimethoxy, 2-methoxyanisole, o,o-dimethyl catechol, 2-dimethoxybenzol, benzene, o-dimethoxy | |
CHEBI:59114 | |
COC1=CC=CC=C1OC |
Specifications
91-16-7 | |
206°C to 207°C | |
Authentic | |
Glass bottle | |
1.5330 to 1.535 | |
MFCD00008357 | |
06, 771 | |
15, 10150 | |
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,methanol,acetone and ether | |
COC1=CC=CC=C1OC | |
138.17 | |
CHEBI:59114 | |
99+% |
1.0800g/mL | |
87°C | |
99% min. (GC) | |
C8H10O2 | |
C6H4(OCH3)2 | |
100g | |
1.08 | |
veratrole, veratrol, pyrocatechol dimethyl ether, o-dimethoxybenzene, catechol dimethyl ether, benzene, 1,2-dimethoxy, 2-methoxyanisole, o,o-dimethyl catechol, 2-dimethoxybenzol, benzene, o-dimethoxy | |
ABDKAPXRBAPSQN-UHFFFAOYSA-N | |
1,2-dimethoxybenzene | |
7043 | |
138.17 | |
Veratrole, 99+% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 202-045-3
RTECSNumber : CZ6475000
TSCA : TSCA
RUO â Research Use Only