Learn More
Veratrole, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1198530
Description
It is useful for organic synthesis of aromatic compounds. Veratrole is relatively electron-rich and thus readily undergoes electrophilic substitution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is useful for organic synthesis of aromatic compounds. Veratrole is relatively electron-rich and thus readily undergoes electrophilic substitution.
Solubility
Soluble in alcohol, diethyl ether, acetone, and methanol. Slightly soluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
Veratrole | |
21°C to 23°C | |
206°C to 207°C | |
C8H10O2 | |
MFCD00008357 | |
1364621 | |
veratrole, veratrol, pyrocatechol dimethyl ether, o-dimethoxybenzene, catechol dimethyl ether, benzene, 1,2-dimethoxy, 2-methoxyanisole, o,o-dimethyl catechol, 2-dimethoxybenzol, benzene, o-dimethoxy | |
ABDKAPXRBAPSQN-UHFFFAOYSA-N | |
1,2-dimethoxybenzene | |
7043 | |
138.17 |
91-16-7 | |
1.084 | |
87°C (188°F) | |
1.533 | |
250 g | |
14,9956 | |
Soluble in alcohol,diethyl ether,acetone,and methanol. Slightly soluble in water. | |
COC1=CC=CC=C1OC | |
138.166 | |
CHEBI:59114 | |
99% |
Chemical Identifiers
91-16-7 | |
138.166 | |
ABDKAPXRBAPSQN-UHFFFAOYSA-N | |
7043 | |
1,2-dimethoxybenzene |
C8H10O2 | |
MFCD00008357 | |
veratrole, veratrol, pyrocatechol dimethyl ether, o-dimethoxybenzene, catechol dimethyl ether, benzene, 1,2-dimethoxy, 2-methoxyanisole, o,o-dimethyl catechol, 2-dimethoxybenzol, benzene, o-dimethoxy | |
CHEBI:59114 | |
COC1=CC=CC=C1OC |
Safety and Handling
GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
EINECSNumber : 202-045-3
RTECSNumber : CZ6475000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only