Please login to your online account to display your discounted pricing

Veratridine, Tocris

Voltage-gated Na+ channel opener

$121.23 - $509.13

Chemical Identifiers

Chemical Name or Material Veratridine
Assay Percent Range >98%
Molecular Formula C36H51NO11
Formula Weight 673.79
Synonym 4α,9-Epoxy-3β-veratroyloxy-5β-cevan-4β,12,14,16β,17,20-hexaol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

291850

 
r & d systems
RDS291850
50mg Each for $509.13

291810

 
r & d systems
RDS291810
10mg Each for $121.23
Description & Specifications

Specifications

Formula Weight 673.79
Quantity 50mg
Chemical Name or Material Veratridine
Synonym 4α,9-Epoxy-3β-veratroyloxy-5β-cevan-4β,12,14,16β,17,20-hexaol
Recommended Storage Store at -20°C
Assay Percent Range >98%
CAS 71-62-5
Molecular Formula C36H51NO11
Safety and Handling

Store at -20°C