Please login to your online account to display your discounted pricing

Venlafaxine hydrochloride, Tocris

Dual serotonin/noradrenalin re-uptake inhibitor

$146.74 - $581.87

Chemical Identifiers

Chemical Name or Material Venlafaxine hydrochloride
Assay Percent Range >99%
Molecular Formula C17H27NO2·HCl
Formula Weight 313.86
Synonym 1-[2-(Dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

291750

 
r & d systems
RDS291750
50mg Each for $581.87

291710

 
r & d systems
RDS291710
10mg Each for $146.74
Description & Specifications

Specifications

Formula Weight 313.86
Quantity 50mg
Molecular Formula C17H27NO2·HCl
Synonym 1-[2-(Dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride
Chemical Name or Material Venlafaxine hydrochloride
Assay Percent Range >99%
CAS 99300-78-4
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature