Please login to your online account to display your discounted pricing

VEGF Receptor 3 Rat anti-Mouse, Clone: AFL4, eBioscience™

Rat Monoclonal Antibody

$114.55 - $346.55

Specifications

Antigen VEGF Receptor 3
Clone AFL4
Host Species Rat
Gene Alias VEGFR3, VEGF R3, VEGF receptor 3, Flt-4, Flt4, vascular endothelial growth factor receptor 3
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013414

 
life technologies
LS14598881
50μg Each for $114.55

5013415

 
life technologies
LS14598882
100μg Each for $230.55

5013416

 
life technologies
LS14598885
500μg Each for $346.55
Description & Specifications

Specifications

Antigen VEGF Receptor 3
Clone AFL4
Host Species Rat
Gene Alias VEGFR3, VEGF R3, VEGF receptor 3, Flt-4, Flt4, vascular endothelial growth factor receptor 3
Species Reactivity Mouse
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The AFL4 monoclonal antibody reacts with the mouse VEGF receptor-3, also known as Flt-4. This 195kDa molecule was identified as an endothelial-specific member of the receptor tyrosine kinase (RTK) family. During early embryogenesis all endothelial cells express VEGFR-3, while in the adult tissues, VEGFR-3 expression disappears from the vascular endothelial cells and is observed only on the lymphatic endothelium. However, VEGFR-3 expression is induced in the adult tissue upon tumor implementation suggesting an important role for this receptor in the tumor angiogenesis. VEGF-C and VEGF-D bind to and activate VEGFR-3. AFL4 is an antagonist mAb.