missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

VEGF Receptor 3 Rat anti-Mouse, Biotin, Clone: AFL4, eBioscience™

Rat Monoclonal Antibody

$117.48 - $381.48

Specifications

Antigen VEGF Receptor 3
Clone AFL4
Host Species Rat
Gene Alias VEGFR3, VEGF R3, VEGF receptor 3, Flt-4, Flt4, vascular endothelial growth factor receptor 3
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012102 Life Technologies
LS13598881
50μg Each for $117.48
5012104 Life Technologies
LS13598885
500μg Each for $381.48
5012103 Life Technologies
LS13598882
100μg Each for $236.28
Specifications

Description

The AFL4 monoclonal antibody reacts with the mouse VEGF receptor-3, also known as Flt-4. This 195kDa molecule was identified as an endothelial-specific member of the receptor tyrosine kinase (RTK) family. During early embryogenesis all endothelial cells express VEGFR-3, while in the adult tissues, VEGFR-3 expression disappears from the vascular endothelial cells and is observed only on the lymphatic endothelium. However, VEGFR-3 expression is induced in the adult tissue upon tumor implementation suggesting an important role for this receptor in the tumor angiogenesis. VEGF-C and VEGF-D bind to and activate VEGFR-3. AFL4 is an antagonist mAb.
Description & Specifications

Specifications

VEGF Receptor 3
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
AFL4
VEGFR3, VEGF R3, VEGF receptor 3, Flt-4, Flt4, vascular endothelial growth factor receptor 3
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL