Please login to your online account to display your discounted pricing

VDM 11, Tocris

Potent, selective anandamide transport inhibitor

$129.05 - $546.19

Chemical Identifiers

Chemical Name or Material VDM 11
Assay Percent Range >98%
Molecular Formula C27H39NO2
Formula Weight 409.61
Synonym (5Z,8Z,11Z,14Z)-N-(4-Hydroxy-2-methylphenyl)-5,8,11,14-eicosatetraenamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

13925

 
r & d systems
RDS13925
5mg Each for $129.05

139225

 
r & d systems
RDS139225
25mg Each for $546.19
Description & Specifications

Specifications

Formula Weight 409.61
Quantity 5mg
Assay Percent Range >98%
CAS 313998-81-1
Synonym (5Z,8Z,11Z,14Z)-N-(4-Hydroxy-2-methylphenyl)-5,8,11,14-eicosatetraenamide
Chemical Name or Material VDM 11
Molecular Formula C27H39NO2
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C