Please login to your online account to display your discounted pricing

VDM 11 (in Tocrisolve 100), Tocris

Potent, selective anandamide transport inhibitor (in water-soluble emulsion)

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material VDM 11 (in Tocrisolve™ 100)
Assay Percent Range >98%
Molecular Formula C27H39NO2
Formula Weight 409.61
Synonym (5Z,8Z,11Z,14Z)-N-(4-Hydroxy-2-methylphenyl)-5,8,11,14-eicosatetraenamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

168625

 
r & d systems
RDS168625
25mg Each for $762.58

16865

  SDS
r & d systems
RDS16865
5mg Each for $181.19
Description & Specifications

Specifications

Formula Weight 409.61
Quantity 25mg
Chemical Name or Material VDM 11 (in Tocrisolve™ 100)
Assay Percent Range >98%
Molecular Formula C27H39NO2
CAS 313998-81-1
Synonym (5Z,8Z,11Z,14Z)-N-(4-Hydroxy-2-methylphenyl)-5,8,11,14-eicosatetraenamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C