Please login to your online account to display your discounted pricing

Alfa Aesar™ Vasoactive Intestinal Peptide (11-28), Human, Porcine, Rat, Ovine

For cellular and molecular biology applications

Specifications

Product Type Vasoactive Intestinal Peptide (11-28), human, porcine, rat, ovine
Cross Reactivity Human, Porcine, Rat, Ovine
Molecular Weight 2143.61
Storage Requirements -30° to -10°C
Sequence Thr-Arg-Leu-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Asn-Ser-Ile-Leu-Asn-NH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66678EXD

 
alfa aesar™
J66678EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Vasoactive Intestinal Peptide (11-28), human, porcine, rat, ovine
Cross Reactivity Human, Porcine, Rat, Ovine
Molecular Weight 2143.61
Storage Requirements -30° to -10°C
Sequence Thr-Arg-Leu-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Asn-Ser-Ile-Leu-Asn-NH2
Format Solid
Common Name VIP (11-28), human, porcine, rat, ovine
Regulatory Status RUO
Purification Method HPLC