Please login to your online account to display your discounted pricing

Alfa Aesar™ Vasoactive Intestinal Peptide (11-28), Human, Porcine, Rat, Ovine


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66678EXD

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66678EXD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Common Name VIP (11-28), human, porcine, rat, ovine
Format Solid
Cross Reactivity Human, Porcine, Rat, Ovine
Molecular Weight 2143.61
Sequence Thr-Arg-Leu-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Asn-Ser-Ile-Leu-Asn-NH2
Purification Method HPLC
Quantity 0.5mg
Product Type Vasoactive Intestinal Peptide (11-28), human, porcine, rat, ovine
Regulatory Status RUO
Storage Requirements -30° to -10°C