Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Vascular Endothelial Growth Factor (VEGF), Rabbit Monoclonal Antibody

Employ part of a comprehensive antibody portfolio with the Thermo Scientific™ Vascular Endothelial Growth Factor, Rabbit Monoclonal Antibody. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

$742.43 - $742.43

Specifications

Antigen Vascular Endothelial Growth Factor (VEGF)
Clone SP28
Applications Immunohistochemistry
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RM9128R7

 
thermo scientific™ lab vision™
RM9128R7
Ready-to-use for IHC (UVLP) 7mL Each for $742.43
Description & Specifications

Specifications

Antigen Vascular Endothelial Growth Factor (VEGF)
Clone SP28
Applications Immunohistochemistry
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Rabbit
Quantity 7mL
Type Primary
Formulation Ready-to-use for IHC (UVLP)
Immunogen A synthetic peptide derived from N-terminal of human VEGF
Isotype IgG
Purification Method Angiosarcoma, VEGF recombinant protein

Employ part of a comprehensive antibody portfolio with the Thermo Scientific™ Vascular Endothelial Growth Factor, Rabbit Monoclonal Antibody. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

VEGF (vascular endothelial growth factor) is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. This antibody reacts with the 165, 189 and 121 amino acid splice variants of VEGF of human, to a lesser extent, mouse and rat.

Host Species: Rabbit

Clone: SP28

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: N-terminus

Immunogen: A synthetic peptide derived from N-terminal of human VEGF

Molecular Weight: 19-22kDa (reduced) or 38-44kDa (nonreduced)

Positive Control: Angiosarcoma, VEGF recombinant protein

Cellular Localization: Cytoplasmic, cell surface in endothelial cells This clone (SP28) is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry

Note:

USA: RUO; Int'l: RUO