missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Vascular Endothelial Growth Factor (VEGF) Ab-7, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Vascular Endothelial Growth Factor (VEGF) Ab-7, Mouse Monoclonal Antibody.

$308.20 - $1,440.59

Specifications

Antigen Vascular Endothelial Growth Factor (VEGF) Ab-7
Clone VG1
Applications Immunohistochemistry (Paraffin),Western Blotting,Immunohistochemistry (Paraffin),Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS1467P Thermo Scientific™ Lab Vision™
MS1467P
Purified Ab with BSA and Azide 1mL Each for $1,351.76
MS1467P0 Thermo Scientific™ Lab Vision™
MS1467P0
Purified Ab with BSA and Azide 100μL Each for $308.20
MS1467P1 Thermo Scientific™ Lab Vision™
MS1467P1
Purified Ab with BSA and Azide 500μL Each for $868.24
MS1467P1ABX Thermo Scientific™ Lab Vision™
MS1467P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $987.33
MS1467PABX Thermo Scientific™ Lab Vision™
MS1467PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,440.59
MS1467R7 Thermo Scientific™ Lab Vision™
MS1467R7
Ready-to-use for IHC (UVLP) 7mL Each for $794.20
Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ Vascular Endothelial Growth Factor (VEGF) Ab-7, Mouse Monoclonal Antibody.

VEGF which is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. It exhibits potent mitogenic and permeability inducing properties specific for the vascular endothelium. Of the four isoforms of VEGF, the smaller two, VEGF165 and VEGF121, are secreted proteins and act as diffusible agents, whereas the larger two (VEGF189 and VEGF206) remain cell associated.

Host Species: Mouse

Clone: VG1

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant VEGF protein

Molecular Weight: 19-22kDa (reduced)

Positive Control: VEGF165 recombinant protein, Angiosarcoma

Cellular Localization: Cytoplasmic, cell surface and extracellular matrix Ab-7 does not neutralize the bioactivity of VEGF.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Vascular Endothelial Growth Factor (VEGF) Ab-7
Immunohistochemistry (Paraffin),Western Blotting,Immunohistochemistry (Paraffin),Western Blotting
Unlabeled
Murine
Angiogenesis
Recombinant VEGF protein
VG1
Monoclonal
Human
RUO
Primary
IgG2a kappa