missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Vascular Endothelial Growth Factor (VEGF) Ab-3, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Vascular Endothelial Growth Factor (VEGF) Ab-3, Mouse Monoclonal Antibody.

$309.92 - $1,326.65

Specifications

Antigen Vascular Endothelial Growth Factor (VEGF) Ab-3
Clone JH121
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS350P View Documents Thermo Scientific™
MS350P
200μg/mL 1mL Each for $1,236.06
 
MS350P0 View Documents Thermo Scientific™
MS350P0
200μg/mL 100μL Each for $309.92
 
MS350P1 View Documents Thermo Scientific™
MS350P1
200μg/mL 500μL Each for $863.84
 
MS350P1ABX View Documents Thermo Scientific™
MS350P1ABX
1mg/mL 100μL Each for $965.49
 
MS350PABX View Documents Thermo Scientific™
MS350PABX
1mg/mL 200μL Each for $1,326.65
 
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Vascular Endothelial Growth Factor (VEGF) Ab-3, Mouse Monoclonal Antibody.

VEGF (vascular endothelial growth factor) is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. It exhibits potent mitogenic and permeability inducing properties specific for the vascular endothelium. Of the four isoforms of VEGF, the smaller two, VEGF165 and VEGF121, are secreted proteins and act as diffusible agents, whereas the larger two (VEGF189 and VEGF206) remain cell associated.

Host Species: Mouse

Clone: JH121

Isotype: IgG1/κ

Species Reactivity: Human and Rabbit. Others not known.

Epitope: Not determined

Immunogen: Recombinant human VEGF121

Molecular Weight: 19-22kDa (reduced)

Positive Control: VEGF165 recombinant protein, Angiosarcoma

Cellular Localization: Cytoplasmic, cell surface and extracellular matrix Ab-3 is excellent for neutralizing the bioactivity of human VEGF.

Recommended for:

  • Neutralizes Bioactivity of VEGF

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Vascular Endothelial Growth Factor (VEGF) Ab-3
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Murine
Angiogenesis
Recombinant human VEGFsubscript121subscript
JH121
Monoclonal
Concentrated
RUO
Primary
IgG2a kappa