Learn More
Vascular Endothelial Growth Factor (VEGF) Ab-3 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Vascular Endothelial Growth Factor (VEGF) Ab-3, Mouse Monoclonal Antibody.
Supplier: Thermo Fisher Scientific MS350P1
Description
VEGF (vascular endothelial growth factor) is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. It exhibits potent mitogenic and permeability inducing properties specific for the vascular endothelium. Of the four isoforms of VEGF, the smaller two, VEGF165 and VEGF121, are secreted proteins and act as diffusible agents, whereas the larger two (VEGF189 and VEGF206) remain cell associated.Host Species: Mouse
Clone: JH121
Isotype: IgG1/κ
Species Reactivity: Human and Rabbit. Others not known.
Epitope: Not determined
Immunogen: Recombinant human VEGF121
Molecular Weight: 19-22kDa (reduced)
Positive Control: VEGF165 recombinant protein, Angiosarcoma
Cellular Localization: Cytoplasmic, cell surface and extracellular matrix Ab-3 is excellent for neutralizing the bioactivity of human VEGF.
Recommended for:
- Neutralizes Bioactivity of VEGF
Note:
USA: RUO; Int'l: RUO
Specifications
Vascular Endothelial Growth Factor (VEGF) Ab-3 | |
Monoclonal | |
200μg/mL | |
Mouse | |
500 μL | |
Angiogenesis | |
Human, Rabbit | |
IgG2a κ |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
JH121 | |
Unconjugated | |
Recombinant human VEGFsubscript121subscript | |
RUO | |
Primary | |
Concentrated |