missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Vascular Endothelial Growth Factor (VEGF) Ab-2, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Vascular Endothelial Growth Factor (VEGF) Ab-2, Mouse Monoclonal Antibody.

$238.63 - $983.00

Specifications

Antigen Vascular Endothelial Growth Factor (VEGF) Ab-2
Clone 5C3.F8
Applications Western Blot
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS290P0 Thermo Scientific™ Lab Vision™
MS290P0
200μg/mL 100μL Each for $238.63
MS290P1 Thermo Scientific™ Lab Vision™
MS290P1
200μg/mL 500μL Each for $663.08
MS290P1ABX Thermo Scientific™ Lab Vision™
MS290P1ABX
1mg/mL 100μL Each for $735.61
MS290PABX Thermo Scientific™ Lab Vision™
MS290PABX
1mg/mL 200μL Each for $983.00
MS290P Thermo Fisher Scientific
MS290P
200μg/mL 1mL Each for $885.47
Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ Vascular Endothelial Growth Factor (VEGF) Ab-2, Mouse Monoclonal Antibody.

VEGF (vascular endothelial growth factor) is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. It exhibits potent mitogenic and permeability inducing properties specific for the vascular endothelium. Of the four isoforms of VEGF, the smaller two, VEGF165 and VEGF121, are secreted proteins and act as diffusible agents, whereas the larger two (VEGF189 and VEGF206) remain cell associated.

Host Species: Mouse

Clone: 5C3.F8

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human VEGF from a glioma

Molecular Weight: 19-22kDa (reduced)

Positive Control: VEGF165 recombinant protein

Cellular Localization: Cytoplasmic, cell surface and extracellular matrix Ab-2 does not neutralize the bioactivity of VEGF.

Recommended for:

  • Western Blotting (Reduced only)
  • Neutralization of VEGF (Not suitable)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Vascular Endothelial Growth Factor (VEGF) Ab-2
Western Blot
Unlabeled
Murine
Angiogenesis
Human VEGF from a glioma
5C3.F8
Monoclonal
Concentrated
RUO
Primary
IgG2a kappa