Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Vascular Endothelial Growth Factor (VEGF) Ab-2, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Vascular Endothelial Growth Factor (VEGF) Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS290P1ABX

 View more versions of this product

Catalog No. MS290P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Vascular Endothelial Growth Factor (VEGF) Ab-2
Applications Western Blot
Classification Monoclonal
Clone 5C3.F8
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Immunogen Human VEGF from a glioma
Isotype IgG2a kappa
Quantity 100μL
Regulatory Status RUO
Research Discipline Angiogenesis
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Provide accurate, reproducible results with the Thermo Scientific™ Vascular Endothelial Growth Factor (VEGF) Ab-2, Mouse Monoclonal Antibody.

VEGF (vascular endothelial growth factor) is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. It exhibits potent mitogenic and permeability inducing properties specific for the vascular endothelium. Of the four isoforms of VEGF, the smaller two, VEGF165 and VEGF121, are secreted proteins and act as diffusible agents, whereas the larger two (VEGF189 and VEGF206) remain cell associated.

Host Species: Mouse

Clone: 5C3.F8

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human VEGF from a glioma

Molecular Weight: 19-22kDa (reduced)

Positive Control: VEGF165 recombinant protein

Cellular Localization: Cytoplasmic, cell surface and extracellular matrix Ab-2 does not neutralize the bioactivity of VEGF.

Recommended for:

  • Western Blotting (Reduced only)
  • Neutralization of VEGF (Not suitable)

Note:

USA: RUO; Int'l: RUO