missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Variamine Blue B Sulfate (=4-Amino-4′-methoxydiphenylamine Sulfate) [for Iron-Titration], TCI America™
$50.42 - $374.31
Chemical Identifiers
CAS | 3169-21-9 |
---|---|
Molecular Formula | C13H16N2O5S |
Molecular Weight (g/mol) | 312.34 |
MDL Number | MFCD00060258 |
InChI Key | BMSZNBNGDJDEOC-UHFFFAOYSA-N |
Synonym | 4-Amino-4′-methoxydiphenylamine Sulfate, N-(4-Methoxyphenyl)-1,4-phenylenediamine Sulfate |
PubChem CID | 165084 |
IUPAC Name | 4-N-(4-methoxyphenyl)benzene-1,4-diamine;sulfuric acid |
SMILES | COC1=CC=C(C=C1)NC2=CC=C(C=C2)N.OS(=O)(=O)O |
Chemical Identifiers
3169-21-9 | |
312.34 | |
BMSZNBNGDJDEOC-UHFFFAOYSA-N | |
165084 | |
COC1=CC=C(C=C1)NC2=CC=C(C=C2)N.OS(=O)(=O)O |
C13H16N2O5S | |
MFCD00060258 | |
4-Amino-4′-methoxydiphenylamine Sulfate, N-(4-Methoxyphenyl)-1,4-phenylenediamine Sulfate | |
4-N-(4-methoxyphenyl)benzene-1,4-diamine;sulfuric acid |
Specifications
3169-21-9 | |
MFCD00060258 | |
BMSZNBNGDJDEOC-UHFFFAOYSA-N | |
4-N-(4-methoxyphenyl)benzene-1,4-diamine;sulfuric acid | |
165084 | |
Crystal-Powder at 20°C | |
Undesignated | |
Variamine Blue B Sulfate (=4-Amino-4′-methoxydiphenylamine Sulfate) [for Iron-Titration] |
C13H16N2O5S | |
4-Amino-4′C-methoxydiphenylamine Sulfate, N-(4-Methoxyphenyl)-1,4-phenylenediamine Sulfate | |
COC1=CC=C(C=C1)NC2=CC=C(C=C2)N.OS(=O)(=O)O | |
312.34 | |
312.34 | |
≥97.0% (N) | |
1g |
Safety and Handling
EINECSNumber : (3)-2801
TSCA : No