missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Variamine Blue B Sulfate (=4-Amino-4′-methoxydiphenylamine Sulfate) 97.0+%, TCI America™

Manufacturer:  TCI America M049525G

 View more versions of this product

Catalog No. M049525G



Specifications

Specifications

Variamine Blue B Sulfate (=4-Amino-4′-methoxydiphenylamine Sulfate) [for Iron-Titration]
C13H16N2O5S
4-Amino-4′-methoxydiphenylamine Sulfate, N-(4-Methoxyphenyl)-1,4-phenylenediamine Sulfate
COC1=CC=C(C=C1)NC2=CC=C(C=C2)N.OS(=O)(=O)O
312.34
312.34
≥97.0% (N)
25g
3169-21-9
MFCD00060258
BMSZNBNGDJDEOC-UHFFFAOYSA-N
4-N-(4-methoxyphenyl)benzene-1,4-diamine;sulfuric acid
165084
Crystal-Powder at 20°C
Undesignated

Chemical Identifiers

3169-21-9
312.34
BMSZNBNGDJDEOC-UHFFFAOYSA-N
165084
COC1=CC=C(C=C1)NC2=CC=C(C=C2)N.OS(=O)(=O)O
C13H16N2O5S
MFCD00060258
4-Amino-4′-methoxydiphenylamine Sulfate, N-(4-Methoxyphenyl)-1,4-phenylenediamine Sulfate
4-N-(4-methoxyphenyl)benzene-1,4-diamine;sulfuric acid
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2801

TSCA : No

Documents