missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Variamine Blue B [Redox Indicator], TCI America™

Manufacturer:  TCI America M04941G

 View more versions of this product

Catalog No. M04941G



Specifications

Specifications

Variamine Blue B [Redox Indicator]
C13H15ClN2O
4-Amino-4′C-methoxydiphenylamine Hydrochloride, N-(4-Methoxyphenyl)-1,4-phenylenediamine Hydrochloride, Variamine Blue B Hydrochloride
COC1=CC=C(C=C1)NC2=CC=C(C=C2)N.Cl
250.726
250.73
≥98.0% (T)
Blue
3566-44-7
MFCD00012989
HPQQXLXIGHOKNZ-UHFFFAOYSA-N
4-N-(4-methoxyphenyl)benzene-1,4-diamine;hydrochloride
77108
Crystal-Powder at 20°C
255°C
1g

Chemical Identifiers

3566-44-7
250.726
HPQQXLXIGHOKNZ-UHFFFAOYSA-N
77108
COC1=CC=C(C=C1)NC2=CC=C(C=C2)N.Cl
C13H15ClN2O
MFCD00012989
4-Amino-4′C-methoxydiphenylamine Hydrochloride, N-(4-Methoxyphenyl)-1,4-phenylenediamine Hydrochloride, Variamine Blue B Hydrochloride
4-N-(4-methoxyphenyl)benzene-1,4-diamine;hydrochloride
Safety and Handling

Safety and Handling

EINECSNumber : (5)-2320

TSCA : Yes

Documents