Variable-Volume Student Micropipets
High accuracy and performance
Manufacturer:  Fisher Scientific AZ600016
 View more versions of this product
Catalog No. S90642A
Description
Description
Durable construction for years of classroom use.
  • Dispense liquids in 1μl increments
  • Easy-to-read digital display
  • Built-in tip ejector
  • Tipcone assembly is autoclavable; easily disassembles for servicing and maintenance
Specifications
Specifications
Yes
Easy-to-read digital display and a built-in tip ejector
1μL
1 to 10μL
Documents