Variable-Volume Student Micropipets
High accuracy and performance
$151.20 - $151.20
Specifications
Autoclavable Yes
Includes Easy-to-read digital display and a built-in tip ejector
Increments 1μL
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
S90642A Fisher Science Education
AZ-60-0016
1 to 10μL Each for $151.20
Specifications
Description
Durable construction for years of classroom use.
  • Dispense liquids in 1μl increments
  • Easy-to-read digital display
  • Built-in tip ejector
  • Tipcone assembly is autoclavable; easily disassembles for servicing and maintenance
Description & Specifications
Specifications
Yes
Easy-to-read digital display and a built-in tip ejector
1μL