Staco Energy™ Variable Transformers
Compact, portable, cord-and-plug type unit
Supplier:  Staco Energy™ 3PN1210B
 View more versions of this product
Catalog No. 09-521-120
Description
Description
Compact, portable, cord-and-plug type unit
  • Three-prong line cord and receptacle
  • Illuminated ON/OFF switch
  • Housed in a rugged, ventilated steel enclosure
  • 50/60Hz
  • Select output voltage from 0-117% line voltage
  • Increase output voltage with a clockwise rotation
Specifications
Specifications
Variable Transformer
6.96 in.
120 V
12 A
5.5 in.
50/60 Hz
10.25 lb.
11.7 cm
Steel
17.68 cm
±5%
120 V, 50/60 Hz
13.9 cm
Three prong plug
4.64 kg