Humboldt Variable-Size Opening Tripods
Manufacturer:  Humboldt HUMH24340
 View more versions of this product
Catalog No. 15-305C
Specifications
Specifications
Flanged Concentric Ring Tripod
8 in.
20.3cm
Sturdy tripods featuring adjustable openings through the use of detachable rings
22.8cm
Cast Iron
3 in.
7.6cm
Black Enamel
9 in.
4 rings
10 Case
Documents