Cole-Parmer™ VapLock™ Activated Media Exhaust Filters
VapLock™ Activated Media Exhaust Filters reduce harmful chemical emissions and odors from solvent containers
Manufacturer:  Cole-Parmer™ 1201980
 View more versions of this product
Catalog No. 13-311-182
Description
Description
Use with indicators to set a replacement schedule
Warranty
None
Specifications
Specifications
Solvent Safety
Activated Carbon with Breakthrough Indicator
Exhaust Filter with Indicator, 75 g activated carbon, 1/4 in. NPT(F); 1/ea
VapLock Solvent Containment Systems
1/4 in. NPT(F)
Documents