Please login to your online account to display your discounted pricing

Alfa Aesar™ Vanillin oxime, 98%

$81.22 - $81.22

Chemical Identifiers

Chemical Name or Material Vanillin oxime
CAS 2874-33-1
Molecular Formula C8H9NO3
Formula Weight 167.17
MDL Number MFCD00019965
Beilstein 1952864
Synonym 4-Hydroxy-3-methoxybenzaldehyde oxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1966706

  SDS
alfa aesar™
L1966706
5g Each for $81.22
Description & Specifications

Specifications

Formula Weight 167.17
Molecular Formula C8H9NO3
CAS 2874-33-1
MDL Number MFCD00019965
Assay 98%
Synonym 4-Hydroxy-3-methoxybenzaldehyde oxime
Quantity 5g
Beilstein 1952864
Melting Point 118° to 122°C
Chemical Name or Material Vanillin oxime