missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanillin, 99%, pure, ACROS Organics™

Manufacturer:  Acros Organics 140820020

 View more versions of this product

Catalog No. AC140820020



Specifications

Specifications

Vanillin
Authentic
White to Yellow
2g
Pure
C8H8O3
15, 10120
MWOOGOJBHIARFG-UHFFFAOYSA-N
4-hydroxy-3-methoxybenzaldehyde
1183
152.15
≥98.5%
1.0600g/mL
Glass Bottle
81°C to 84°C
121-33-5
99%
08, 247
vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
COC1=C(C=CC(=C1)C=O)O
152.149
CHEBI:18346
Crystalline Powder

Chemical Identifiers

121-33-5
152.149
vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
CHEBI:18346
COC1=C(C=CC(=C1)C=O)O
C8H8O3
MWOOGOJBHIARFG-UHFFFAOYSA-N
1183
4-hydroxy-3-methoxybenzaldehyde
Safety and Handling

Safety and Handling

Warning

SDS Documents