missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanillin, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1116936

 View more versions of this product

Catalog No. AAA1116936



Description

Description

Applications
Vanillin is a flavoring agent used in sweet foods, ice cream and chocolate. It is used in perfumes as well as to control the unpleasant odors in medicines. It acts as a chemical intermediate in the preparation of pharmaceuticals. In thin layer chromatography, it is used as a general-purpose stain for developing TLC plates to enable in visualizing components of a reaction mixture. Similarly, vanillin hydro chloride staining can be used to visualize the localization of tannins in cells.

Solubility
Soluble in water, diethyl ether, acetone, methanol, benzene, petroleum ether, chloroform, carbon disulfide, glacial acetic acid, pyridine and in aqueous solutions of alkali hydroxides.

Notes
Moisture and light sensitive. Incompatible with oxidizing agents, alkalis and reducing agents.
Specifications

Specifications

Vanillin
500g
99%
MFCD00006942
14,9932
MWOOGOJBHIARFG-UHFFFAOYSA-N
4-hydroxy-3-methoxybenzaldehyde
1183
152.15
1.06g/mL
121-33-5
C8H8O3
472792
vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
COC1=C(C=CC(=C1)C=O)O
152.149
CHEBI:18346
99%

Chemical Identifiers

121-33-5
152.149
MWOOGOJBHIARFG-UHFFFAOYSA-N
1183
4-hydroxy-3-methoxybenzaldehyde
C8H8O3
MFCD00006942
vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
CHEBI:18346
COC1=C(C=CC(=C1)C=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332

GHS P Statement
P261-P264-P270-P271-P304+P340-P301+P312-P312-P330-P501a

Warning

RTECSNumber : YW5775000

TSCA : TSCA

SDS Documents