missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Vanillic acid, 98%

Manufacturer:  Alfa Aesar™ A1207436

 View more versions of this product

Catalog No. AAA1207436


 

Description

Description

Applications
Vanillic acid is used as a flavoring agent in food. It acts as an intermediate in the production of vanillin from ferulic acid. Further, it is used in wine and vinegar.

Solubility
Soluble in water, alcohol and ether.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Vanillic acid
98%
C8H8O4
2208364
4-Hydroxy-3-methoxybenzoic acid
COC1=C(C=CC(=C1)C(=O)O)O
168.148
CHEBI:30816
210°C to 218°C
121-34-6
99%
MFCD00002551
14,9931
WKOLLVMJNQIZCI-UHFFFAOYSA-N
4-hydroxy-3-methoxybenzoic acid
8468
168.15
500g

Chemical Identifiers

121-34-6
168.148
WKOLLVMJNQIZCI-UHFFFAOYSA-N
8468
C8H8O4
MFCD00002551
4-Hydroxy-3-methoxybenzoic acid
CHEBI:30816
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : YW5300000

TSCA : TSCA

SDS Documents