Please login to your online account to display your discounted pricing

Alfa Aesar™ Vancomycin hydrochloride, 50mg/mL in distilled water, sterile-filtered

Chemical Identifiers

Chemical Name or Material Vancomycin hydrochloride
CAS 1404-93-9
Name Note 50mg/mL in distilled water, sterile-filtered
MDL Number MFCD03613611
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ672518EQ

 
alfa aesar™
J672518EQ
5 x 1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Vancomycin hydrochloride
CAS 1404-93-9
Physical Form Liquid
MDL Number MFCD03613611
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H317
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 -30° to -10°C
RTECS Number YW4380000
Name Note 50mg/mL in distilled water, sterile-filtered
Quantity 5 x 1mL
Safety and Handling

Warning

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H317
May cause an allergic skin reaction.

-30° to -10°C