Please login to your online account to display your discounted pricing

Vanadyl phthalocyanine, 85%, pract., ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  AC422491000

 View more versions of this product

Catalog No. AC422491000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥80%
Physical Form Crystalline Powder or Crystals and Chunks
Formula Weight 579.46
Quantity 100g
Color White
CAS 13930-88-6
Assay Percent Range 85%
Infrared Spectrum Authentic
Health Hazard 2 GHS P Statement Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.
UV_Vis 80% min.
Molecular Formula C32H16N8OV
Health Hazard 1 Warning
Chemical Name or Material Vanadyl phthalocyanine
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF

Specifications

Chemical Name or Material Vanadyl phthalocyanine
Assay ≥80%
Assay Percent Range 85%
Molecular Formula C32H16N8OV
Formula Weight 579.46
Accessories

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF

GHS P Statement Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.

SDS & Safety Documents