Please login to your online account to display your discounted pricing

Vanadyl(IV) sulfate hydrate, 17-23% V, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC214840250

 View more versions of this product

Catalog No. AC214840250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 163
Quantity 25g
Color White to slightly yellow
Assay 17 to 23% (V)
RTECS Number YW1840000
Chemical Name or Material Vanadyl(IV) sulfate hydrate
Iron (Fe) 600ppm max.
Name Note 17-23% V
Packaging Glass bottle
Potassium (K) 500ppm max.
Potassium Oxide 2000ppm max.
Sodium (Na) 500ppm max.
Sodium Oxide 4000ppm max.
Molecular Formula O5SV·xH2O
Linear Formula VOSO4·xH2O
MDL Number MFCD00149933
CAS 12439-96-2
Synonym Oxysulfato vanadium (IV) hydrate
Health Hazard 1 GHS Signal Word: Warning
Additional Information Typical moisture content: 35% (ca. 5 moles water)
Health Hazard 2 GHS P Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Vanadyl(IV) sulfate hydrate
Name Note 17-23% V
CAS 12439-96-2
Assay 17 to 23% (V)
Molecular Formula O5SV·xH2O
Linear Formula VOSO4·xH2O
Formula Weight 163
MDL Number MFCD00149933
Synonym Oxysulfato vanadium (IV) hydrate
Accessories

GHS Signal Word: Warning

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

SDS & Safety Documents