Please login to your online account to display your discounted pricing

Vanadyl(IV) acetylacetonate, 99%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  199592500

 View more versions of this product

Catalog No. AC199592500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 265.15
Physical Form Crystalline Powder
Quantity 250g
Color Green to blue-green
TSCA TSCA
EINECS Number 221-590-8
Fieser 02,456; 03,331; 11,47; 13,54; 15,57; 15,186
Infrared Spectrum Authentic
Health Hazard 1 Warning
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
Molecular Formula C10H14O5V
Linear Formula (CH3COCH=C(CH3)O)2V=O
Chemical Name or Material Vanadyl(IV) acetylacetonate
Assay Percent Range 99%
CAS 3153-26-2
MDL Number MFCD00000032
Name Note 99%

Specifications

Chemical Name or Material Vanadyl(IV) acetylacetonate
Name Note 99%
CAS 3153-26-2
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H14O5V
Linear Formula (CH3COCH=C(CH3)O)2V=O
Formula Weight 265.15
MDL Number MFCD00000032
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.

SDS & Safety Documents