missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Vanadomolybdate Solution, in Dilute Perchloric Acid, LabChem™
Manufacturer: Lab Chem LC266202
Specifications
Vanadomolybdate Solution | |
88.2,8.8,2,1 | |
(NH4)6Mo7O24·4H2O | |
1235.86 | |
Passes Test | |
Soluble in water | |
Gaseous Ammonia; Thermal decomposition generates: Corrosive vapors | |
1L |
in Dilute Perchloric Acid | |
7601-90-3,12054-85-2,7803-55-6 | |
dihydrogen monoxide | |
Liquid | |
Poly Bottle | |
Room Temperature | |
Yellow |