missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Vanadium wire, 1.0mm (0.04 in.) dia., stress relieved, 99.7% (metals basis)

Manufacturer:  Alfa Aesar™ 14359BY

 View more versions of this product

Catalog No. AA14359BY



Specifications

Specifications

Vanadium wire
100cm
7440-62-2
V
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
[V]
Odorless
23990
Wire
≈4.8g/m
1.0mm (0.04 in.) dia., Stress relieved
(metals basis)
MFCD00011453
LEONUFNNVUYDNQ-UHFFFAOYSA-N
vanadium
50.941
CHEBI:35170
99.7%

Chemical Identifiers

7440-62-2
50.941
LEONUFNNVUYDNQ-UHFFFAOYSA-N
23990
vanadium
V
MFCD00011453
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
CHEBI:35170
[V]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents