missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Vanadium wire, 0.25mm (0.01 in.) dia., 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 44105G1

 View more versions of this product

Catalog No. AA44105G1



Specifications

Specifications

Vanadium wire
1m
7440-62-2
V
00j9j9xkde
[V]
Odorless
23990
Wire, ≈0.3g/m
≈0.3g/m
0.25mm (0.01 in.) dia.
(metals basis)
MFCD00011453
LEONUFNNVUYDNQ-UHFFFAOYSA-N
vanadium
50.941
CHEBI:35170
99.8%

Chemical Identifiers

7440-62-2
50.941
LEONUFNNVUYDNQ-UHFFFAOYSA-N
23990
vanadium
V
MFCD00011453
00j9j9xkde
CHEBI:35170
[V]
Safety and Handling

Safety and Handling

EINECSNumber : 231-171-1

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents