missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanadium wire, 0.127mm (0.005in) dia, 99.8% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 011483G5

 View more versions of this product

Catalog No. AA11483G5



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Vanadium wire
7440-62-2
V
5 m
LEONUFNNVUYDNQ-UHFFFAOYSA-N
vanadium
23990
99.8%
≈0.08g/m
0.127mm (0.005 in.) dia.
(metals basis)
MFCD00011453
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
[V]
50.94
CHEBI:35170
Odorless
Wire

Chemical Identifiers

7440-62-2
50.94
LEONUFNNVUYDNQ-UHFFFAOYSA-N
23990
vanadium
V
MFCD00011453
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
CHEBI:35170
[V]
Safety and Handling

Safety and Handling

EINECSNumber : 231-171-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only