missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Vanadium(V) triisopropoxide oxide, 96%

Manufacturer:  Alfa Aesar™ 8979814

 View more versions of this product

Catalog No. AA8979814



Specifications

Specifications

Vanadium (V) triisopropoxide oxide
C9H24O4V
MFCD00015017
vanadium v oxytriisopropoxide, oxotris propan-2-olato vanadium, vanadium v triisopropoxide oxide, oxotri isopropoxo vanadium, vtip, vo oipr 3, vanadium triisopropoxy oxide, triisopropoxy oxo vanadium, vanadium v tri-isopropoxy oxide, dsggjxauqhkogq-uhfffaoysa-n
CC(C)O.CC(C)O.CC(C)O.O=[V]
247.228
242.2
96%
Moisture Sensitive
Orange-Yellow
25g
5588-84-1
VO[CHO(CH3)2]3
UN1993
JOUSPCDMLWUHSO-UHFFFAOYSA-N
oxovanadium;propan-2-ol
79702
Liquid
1.04g/mL
81°C to 83°C (2mmHg)
-12°C
45°C (113°F)

Chemical Identifiers

5588-84-1
247.228
JOUSPCDMLWUHSO-UHFFFAOYSA-N
79702
CC(C)O.CC(C)O.CC(C)O.O=[V]
C9H24O4V
MFCD00015017
vanadium v oxytriisopropoxide, oxotris propan-2-olato vanadium, vanadium v triisopropoxide oxide, oxotri isopropoxo vanadium, vtip, vo oipr 3, vanadium triisopropoxy oxide, triisopropoxy oxo vanadium, vanadium v tri-isopropoxy oxide, dsggjxauqhkogq-uhfffaoysa-n
oxovanadium;propan-2-ol
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

SDS Documents