Please login to your online account to display your discounted pricing

Alfa Aesar™ Vanadium(V) triisopropoxide oxide, 96%

$215.47 - $215.47

Chemical Identifiers

Chemical Name or Material Vanadium (V) triisopropoxide oxide
CAS 5588-84-1
Molecular Formula C9H21O4V
Linear Formula VO[CHO(CH3)2]3
Formula Weight 242.2
MDL Number MFCD00015017
UN Number UN1993
Synonym Isopropyl orthovanadate; Vanadium(V) oxytriisopropoxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA8979814

  SDS
alfa aesar™
8979814
25g Each for $215.47

AA8979803

  SDS
alfa aesar™
8979803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 242.2
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point −12°C
Sensitivity Moisture Sensitive
UN Number UN1993
Flash Point 45°C (113°F)
Linear Formula VO[CHO(CH3)2]3
Health Hazard 1 Warning
Boiling Point 81° to 83°C (2mmHg)
Color Pale yellow-orange
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 25g
Synonym Isopropyl orthovanadate; Vanadium(V) oxytriisopropoxide
Chemical Name or Material Vanadium (V) triisopropoxide oxide
Assay 96%
Molecular Formula C9H21O4V
MDL Number MFCD00015017
Density 1.04g/mL
Physical Form Liquid
TSCA TSCA
CAS 5588-84-1
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.