Learn More
Alfa Aesar™ Vanadium(V) trifluoride oxide, 99%
Manufacturer: Alfa Aesar™ 1412806
Specifications
Vanadium(V) trifluoride oxide | |
F3H3OV | |
UN3290 | |
HHQFLEDKAVLHOM-UHFFFAOYSA-N | |
oxovanadium;trihydrofluoride | |
123322 | |
Powder | |
Moisture Sensitive | |
5g |
13709-31-4 | |
MFCD00042555 | |
trifluorooxovanadium, trifluoro oxo vanadium, vanadium oxytrifluoride, vof3, vanadium v trifluoride oxide, vanadium v oxyfluoride, vanadium v oxyfluoride, vanadium,trifluorooxo-, t-4-9ci | |
O=[V].F.F.F | |
126.96 | |
123.94 | |
99% | |
300°C |
Chemical Identifiers
13709-31-4 | |
126.96 | |
HHQFLEDKAVLHOM-UHFFFAOYSA-N | |
123322 | |
O=[V].F.F.F |
F3H3OV | |
MFCD00042555 | |
trifluorooxovanadium, trifluoro oxo vanadium, vanadium oxytrifluoride, vof3, vanadium v trifluoride oxide, vanadium v oxyfluoride, vanadium v oxyfluoride, vanadium,trifluorooxo-, t-4-9ci | |
oxovanadium;trihydrofluoride |
Safety and Handling
GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLID, CORROSIVE, INORGANIC, N.O.S.