missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Vanadium(V) trifluoride oxide, 99%

Manufacturer:  Alfa Aesar™ 1412814

 View more versions of this product

Catalog No. AA1412814



Specifications

Specifications

Vanadium(V) trifluoride oxide
F3H3OV
UN3290
HHQFLEDKAVLHOM-UHFFFAOYSA-N
oxovanadium;trihydrofluoride
123322
Powder
Moisture Sensitive
25g
13709-31-4
MFCD00042555
trifluorooxovanadium, trifluoro oxo vanadium, vanadium oxytrifluoride, vof3, vanadium v trifluoride oxide, vanadium v oxyfluoride, vanadium v oxyfluoride, vanadium,trifluorooxo-, t-4-9ci
O=[V].F.F.F
126.96
123.94
99%
300°C

Chemical Identifiers

13709-31-4
126.96
HHQFLEDKAVLHOM-UHFFFAOYSA-N
123322
O=[V].F.F.F
F3H3OV
MFCD00042555
trifluorooxovanadium, trifluoro oxo vanadium, vanadium oxytrifluoride, vof3, vanadium v trifluoride oxide, vanadium v oxyfluoride, vanadium v oxyfluoride, vanadium,trifluorooxo-, t-4-9ci
oxovanadium;trihydrofluoride
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLID, CORROSIVE, INORGANIC, N.O.S.

Documents