Please login to your online account to display your discounted pricing

Alfa Aesar™ Vanadium(V) trichloride oxide, V+5 28.5% min

$16.01 - $16.01

Chemical Identifiers

Chemical Name or Material Vanadium (V) trichloride oxide
CAS 7727-18-6
Molecular Formula VOCl3
Formula Weight 173.3
MDL Number MFCD00011459
UN Number UN2443
Merck Index 14,9927
Synonym Vanadium oxytrichloride; Vanadyl trichloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA8111809

  SDS
alfa aesar™
8111809*
10g Each for $16.01
Description & Specifications

Specifications

Formula Weight 173.3
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Vanadium (V) trichloride oxide
TSCA TSCA
Odor Chlorine-like
Physical Form Liquid
Sensitivity Moisture Sensitive
Molecular Formula VOCl3
MDL Number MFCD00011459
CAS 7727-18-6
UN Number UN2443
Merck Index 14,9927
Synonym Vanadium oxytrichloride; Vanadyl trichloride
Boiling Point 126° to 127°C
Color Yellow
Melting Point −77°C
Quantity 10g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Concentration V+5 28.5% min
Density 1.829g/mL
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: VANADIUM OXYTRICHLORIDE
RTECS Number YW2975000
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.