Learn More
Vanadium(V) oxytriethoxide, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 380010050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Vanadium(V) oxytriethoxide | |
1.1390g/mL | |
71°C | |
95% | |
C6H15O4V | |
(CH3CH2O)3V=O | |
5mL | |
triethyl vanadate, vanadium v oxytriethoxide, triethoxyoxovanadium, triethoxo-oxovanadium, vanadium, triethoxyoxo-, t-4, ethylorthovanadate, triethoxy oxo vanadium, iohmvqgdvucilm-uhfffaoysa-n | |
CCO.CCO.CCO.O=[V] | |
202.12 | |
202.12 |
1686-22-2 | |
125.0°C to 128.0°C | |
Authentic | |
Glass bottle | |
1.5080 to 1.5120 | |
MFCD00799170 | |
1.139 | |
IDIDIJSLBFQEKY-UHFFFAOYSA-N | |
ethanol;oxovanadium | |
74312 | |
95% |
Chemical Identifiers
1686-22-2 | |
202.12 | |
IDIDIJSLBFQEKY-UHFFFAOYSA-N | |
74312 | |
CCO.CCO.CCO.O=[V] |
C6H15O4V | |
MFCD00799170 | |
triethyl vanadate, vanadium v oxytriethoxide, triethoxyoxovanadium, triethoxo-oxovanadium, vanadium, triethoxyoxo-, t-4, ethylorthovanadate, triethoxy oxo vanadium, iohmvqgdvucilm-uhfffaoysa-n | |
ethanol;oxovanadium |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 216-870-1
RUO â Research Use Only