Please login to your online account to display your discounted pricing

Vanadium(V) oxide, Puratronic™, 99.99% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Vanadium(V) oxide
CAS 1314-62-1
Name Note Puratronic™
Molecular Formula V2O5
Formula Weight 181.88
MDL Number MFCD00011457
UN Number UN2862
Merck Index 14,9921
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1090418

  SDS
alfa aesar
1090418
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.88
Quantity 50g
Assay 99.99%
Boiling Point 1750°C (Dec.)
Density 3.357
CAS 1314-62-1
Name Note Puratronic™
Assay Percent Notes (metals basis)
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM PENTOXIDE
Molecular Formula V2O5
RTECS Number YW2125000
Mesh Size −22 Mesh
Merck Index 14,9921
Color Orange
MDL Number MFCD00011457
TSCA TSCA
Chemical Name or Material Vanadium(V) oxide
Physical Form Powder
Melting Point 690°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H372-H341-H361-H302-H332-H335
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Odorless
UN Number UN2862
Safety and Handling

Danger

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H372-H341-H361-H302-H332-H335
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.