Learn More
Vanadium(V) oxide, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 450160010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1750.0°C | |
690.0°C | |
1 kg | |
O5V2 | |
MFCD00011457 | |
vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride | |
GNTDGMZSJNCJKK-UHFFFAOYSA-N | |
[(dioxovanadio)oxy]dioxovanadium | |
14814 | |
181.88 | |
Plastic bottle | |
3.3500g/mL |
Vanadium(V) oxide | |
1314-62-1 | |
99% min | |
V2O5 | |
15,10107 | |
Solubility in water: 8g/L. Other solubilities: soluble in acids and alkalies, insoluble in alcohol | |
O=[V](=O)O[V](=O)=O | |
181.88 | |
CHEBI:30045 | |
≥99% | |
3.35 |
Chemical Identifiers
1314-62-1 | |
181.88 | |
GNTDGMZSJNCJKK-UHFFFAOYSA-N | |
14814 | |
O=[V](=O)O[V](=O)=O |
O5V2 | |
MFCD00011457 | |
vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride | |
CHEBI:30045 |
Safety and Handling
GHS H Statement
Suspected of damaging the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes damage to organs through prolonged or re
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
GHS Signal Word: Danger
EINECSNumber : 215-239-8
RUO – Research Use Only